missing translation for 'onlineSavingsMsg'
Learn More

Nitrofurantoin, TRC

Product Code. 30135445
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135445 25 g 25g
30040499 1 g 1g
17909869 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135445 Supplier Toronto Research Chemicals Supplier No. TRCN49385025G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-20-9
Molecular Formula C8 H6 N4 O5
Molecular Weight (g/mol) 238.16
Synonym Nitrofurantoin, 1-[[(5-Nitro-2-furanyl)methylene]amino]-2,4-imidazolidinedione, 1-(5-Nitro-2-furfurylidenamino)hydantoin, 1-(5-Nitro-2-furfurylideneamino)hydantoin, 1-[(5-Nitrofurfurylidene)amino]hydantoin, 5-Nitrofurantoin, Berkfurin, Chemiofuran, Cyantin, Cystit, FuaMed, Fur-ren, Furachel, Furadantin, Furadantine MC, Furadantoin, Furadoin, Furadoine, Furadonin, Furadonine, Furadontin, Furalan, Furantoin, Furantoina, Furatoin, Furina, Furobactina, Furophen T-Caps, Ituran, Macrobid, Macrodantin, N-(5-Nitro-2-furfurylidene)-1-aminohydantoin, N-(5-Nitro-2-furfurylidine)-1-aminohydantoin, N-(5-Nitrofurfurylidene)-1-aminohydantoin, NSC 2107, NSC 44150, Nifurantin, Nitoin, Novofuran, Orafuran, Parfuran, Trantoin, Urantoin, Urizept, Uro-Tablinen, Urodin, Urofurin, Urolong, Welfurin, Zoofurin
IUPAC Name 1-[(E)-(5-nitrofuran-2-yl)methylideneamino]imidazolidine-2,4-dione
SMILES [O-][N+](=O)c1oc(\C=N\N2CC(=O)NC2=O)cc1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C8H6N4O5/c13-6-4-11(8(14)10-6)9-3-5-1-2-7(17-5)12(15)16/h1-3H,4H2,(H,10,13,14)/b9-3+
Formula Weight 238.0338
Percent Purity >95%
Chemical Name or Material Nitrofurantoin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.