missing translation for 'onlineSavingsMsg'
Learn More

Nitrofurate, TRC

Product Code. 30135777
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135777 10 g 10g
30008421 250 mg 250mg
30003803 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135777 Supplier Toronto Research Chemicals Supplier No. TRCN49595510G
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 645-12-5
Molecular Formula C5H3NO5
Molecular Weight (g/mol) 157.08
Synonym Nitrofuroic Acid, 5-Nitro-2-furancarboxylic Acid, 5-Nitro-2-furoic Acid, 5-Nitro-2-furylcarboxylic Acid, 5-Nitrofuroic Acid, 5-Nitropyromucic Acid, NSC 6452
IUPAC Name 5-nitrofuran-2-carboxylic acid
SMILES C1=C(C(=O)O)OC(=C1)[N+](=O)[O-]

Specifications

Quantity 10 g
InChI Formula InChI=1S/C5H3NO5/c7-5(8)3-1-2-4(11-3)6(9)10/h1-2H,(H,7,8)
Formula Weight 157.0011
Chemical Name or Material Nitrofurate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.