missing translation for 'onlineSavingsMsg'
Learn More

Nitrofurazone, MedChemExpress

Product Code. 16462099
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16462099 1 g 1g
16410838 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16462099 Supplier Medchem Express Supplier No. HYB0226
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Nitrofurazone (Nitrofural) is a bactericidal compound used as an antibiotic most commonly in the form of ointments.

Nitrofurazone (Nitrofural) is a bactericidal compound used as an antibiotic most commonly in the form of ointments.

Specifications

Chemical Name or Material Nitrofurazone
CAS 59-87-0
Color Yellow
Purity Grade Notes Research
Molecular Formula C6H6N4O4
Quantity 1 g
Synonym Nitrofural
Solubility Information DMSO : ≥ 155 mg/mL (782.28 mM)
SMILES O=C(N/N=C/C1=CC=C([N+]([O-])=O)O1)N
Molecular Weight (g/mol) 198.14
Formula Weight 198.14
Percent Purity 98.04%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.