missing translation for 'onlineSavingsMsg'
Learn More

Nitromethane, 96%, for spectroscopy

Product Code. 10792971 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
2.5 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10792971 2.5 L Glass bottle 2.5 L
10448920 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 10792971 Supplier Thermo Scientific Acros Supplier No. 167860025
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

CAS: 75-52-5 | CH3NO2 | 61.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-52-5
Molecular Formula CH3NO2
Molecular Weight (g/mol) 61.04
MDL Number MFCD00007400
InChI Key LYGJENNIWJXYER-UHFFFAOYSA-N
Synonym methane, nitro, nitrocarbol, nitrometan, nitrometan polish, nitro-methane, unii-ru5wg8c3f4, ccris 1205, hsdb 106, meno2, ru5wg8c3f4
PubChem CID 6375
ChEBI CHEBI:77701
IUPAC Name nitromethane
SMILES C[N+]([O-])=O

Specifications

Absorbance 0.01 max. at 420nm, 0.05 max. at 405nm, 0.1 max. at 400nm, 0.2 max. at 395nm, 0.5 max. at 386nm, 1 max. at 380nm
CAS Min % 95.0
CAS Max % 100.0
Melting Point -29°C
Density 1.1200g/mL
Boiling Point 100°C to 102°C
Flash Point 35°C
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula CH3NO2
Refractive Index 1.3805 to 1.3825
Quantity 2.5 L
Beilstein 01, 74
Fieser 01,739; 04,357; 09,323; 13,199
Merck Index 15, 6698
Specific Gravity 1.12
Solubility Information Solubility in water: 95g/L (20°C). Other solubilities: soluble in alcohol,ether and dimethylformamide
Viscosity 0.61 mPa.s (25°C)
Formula Weight 61.04
Percent Purity 96%
Grade Spectroscopy
Physical Form Liquid
Chemical Name or Material Nitromethane, 96%, spectrophotometric grade
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Nitromethane
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.