missing translation for 'onlineSavingsMsg'
Learn More

Nitromethane-d3, for NMR, 99.5 atom % D

Product Code. 16348216 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16348216 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 16348216 Supplier Thermo Scientific Acros Supplier No. 464230050
  • ONLINE EXCLUSIVE PRICE
    107.00 €
    103.00 € / 5mL
    Save 4.00 €
    3% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

CAS: 13031-32-8 | CH3NO2 | 64.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Density 1.1800 g/mL
Flash Point 35°C
Refractive Index 1.3795
Specific Gravity 1.18
Quantity 5 mL
CAS 13031-32-8
Boiling Point 100.0°C to 102.0°C
Molecular Formula CH3NO2
Linear Formula CD3NO2
MDL Number MFCD00044214
InChI Key LYGJENNIWJXYER-FIBGUPNXSA-N
SMILES [2H]C([2H])([2H])[N+]([O-])=O
IUPAC Name nitro(²H₃)methane
Molecular Weight (g/mol) 64.06
Formula Weight 64.06
Grade NMR
Chemical Name or Material Nitromethane-d3
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.