missing translation for 'onlineSavingsMsg'
Learn More

Nitromethane-d3 (Reagent grade), CDN

Product Code. 30316838
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30316838 10 g 10g
30309375 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30316838 Supplier CDN Supplier No. CDND011810G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nitromethane-D3
CAS 13031-32-8
Molecular Formula C D3 N O2
Quantity 10 g
InChI Formula InChI=1 S/CH3NO2/c1-2(3)4/h1H3/i1D3
SMILES [2 H]C([2 H])([2 H])[N+](=O)[O-]
IUPAC Name trideuterio(nitro)methane
Molecular Weight (g/mol) 64.058
Formula Weight 64.035 g/mol
Percent Purity 99 atom % D, min 99% Chemical Purity
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.