missing translation for 'onlineSavingsMsg'
Learn More

Nitronium tetrafluoroborate, 0.3 to 0.5M soln. in sulfolane

Product Code. 15478765
Change view
Click to view available options
Quantity:
25 mL
200 mL
Unit Size:
200mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15478765 200 mL 200mL
15468765 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15478765 Supplier Thermo Scientific Alfa Aesar Supplier No. H34069.AH
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13826-86-3 | BF4NO2 | 132.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13826-86-3
Molecular Formula BF4NO2
Molecular Weight (g/mol) 132.81
MDL Number MFCD00011432
InChI Key XYRQURYIJKJVSF-UHFFFAOYSA-N
Synonym nitronium tetrafluoroborate, nitronium ion tetrafluoroborate, nitronium tetrafluoroboron, nitroniumtetrafluoroborate, nitronium.tetrafluoroborate, nitronium tetrafluoroborate, 0.3-0.5m solution in sulfolane
PubChem CID 11073463
IUPAC Name nitrous acid; tetrafluoroboranuide
SMILES [O-][NH+]=O.F[B-](F)(F)F

Specifications

Quantity 200 mL
Linear Formula NO2BF4
UN Number UN3265
Formula Weight 132.81
Concentration or Composition (by Analyte or Components) 0.3 to 0.5M soln. in sulfolane
Sensitivity Moisture sensitive
Density 1.268
Chemical Name or Material Nitronium tetrafluoroborate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.