missing translation for 'onlineSavingsMsg'
Learn More

Nitronium Tetrafluoroborate (0.5 M solution in Sulfolane), TRC

Product Code. 30483097
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30483097 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30483097 Supplier Toronto Research Chemicals Supplier No. TRCN5978805G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nitronium Tetrafluoroborate
CAS 13826-86-3
Molecular Formula BF4NO2
Quantity 5 g
Synonym Nitryl Fluoborate, Nitryl tetrafluoroborate, Nitronium Fluoroborate, Nitronium Tetrafluoroborate, Nitronium Tetrafluoroborate, Nitryl Tetrafluoroborate, Nitryl Tetrafluoroborate
InChI Formula InChI=1S/BF4.NO2/c2-1(3,4)5;2-1-3/q-1;+1
SMILES F[B-](F)(F)F.O=[N+]=O
IUPAC Name nitronium;tetrafluoroborate
Molecular Weight (g/mol) 132.81
Formula Weight 132.9958
Concentration or Composition (by Analyte or Components) 0.5 M
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.