missing translation for 'onlineSavingsMsg'
Learn More

Nitronium tetrafluoroborate, 0.5M solution in sulfolane, AcroSeal™

Product Code. 10311140 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10311140 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10311140 Supplier Thermo Scientific Acros Supplier No. 429071000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 13826-86-3 | BF4NO2 | 132.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 13826-86-3, 126-33-0
Color Colorless through Yellow
CAS Min % 5.03, 94.24
CAS Max % 5.76, 94.97
Physical Form Solution
Quantity 100 mL
Molecular Formula BF4NO2
Linear Formula NO2BF4
MDL Number MFCD00011432
Synonym nitronium tetrafluoroborate, nitronium ion tetrafluoroborate, nitronium tetrafluoroboron, nitroniumtetrafluoroborate, nitronium.tetrafluoroborate, nitronium tetrafluoroborate, 0.3-0.5m solution in sulfolane
Solubility Information Solubility in water: reacts
InChI Key XYRQURYIJKJVSF-UHFFFAOYSA-N
SMILES [O-][NH+]=O.F[B-](F)(F)F
IUPAC Name nitronium;tetrafluoroborate
Molecular Weight (g/mol) 132.81
PubChem CID 11073463
Formula Weight 132.81
Packaging AcroSeal™ Glass Bottle
Specific Gravity 1.268
Density 1.2680g/mL
Chemical Name or Material Nitronium tetrafluoroborate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Nitronium tetrafluoroborate, 0.5 M solution in sulfolane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.