missing translation for 'onlineSavingsMsg'
Learn More

Nitrosobis(2-hydroxyethyl)amine, TRC

Product Code. 30041267
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30041267 1 g 1g
18010730 100 mg 100mg
18022620 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30041267 Supplier Toronto Research Chemicals Supplier No. TRCN5252001G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1116-54-7
Molecular Formula C4 H10 N2 O3
Molecular Weight (g/mol) 134.13
Synonym 2,2'-(Nitrosoimino)bisethanol, Ethanol, 2,2'-nitrosiminodi- (6CI,7CI,8CI), Diethanolnitrosamine, N,N-Diethanolnitrosamine, N-Nitrosobis(2-hydroxyethyl)amine, N-Nitrosodiethanolamine, NDELA, Nitrosodiethanolamine
IUPAC Name N,N-bis(2-hydroxyethyl)nitrous amide
SMILES OCCN(CCO)N=O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C4H10N2O3/c7-3-1-6(5-9)2-4-8/h7-8H,1-4H2
Formula Weight 134.0691
Percent Purity >95%
Chemical Name or Material N-Nitroso-diethanolamine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.