missing translation for 'onlineSavingsMsg'
Learn More

Nitrosonium tetrafluoroborate, 98%

Product Code. p-7022854
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
This item is not returnable. View return policy

Product Code. 11468730

Brand: Thermo Scientific Alfa Aesar A15806.04

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14635-75-7 | BF4HNO2 | 133.82 g/mol

Nitrosonium tetrafluoroborate is a nitrosating agent and oxidizer used in organic synthesis. It is also used to convert arylamines directly to diazonium tetrafluoroborates, which undergo the Balz-Schiemann reaction in situ to give high yields of aryl fluorides. Further, it is used as a catalyst for iodination of aromatics by iodide ion with air as oxidant, conversion of secondary amines to nitrosamines and in deamination of primary amides. It is involved in the conversion of arylhydrazines to azides. In addition to this, it reacts with ferrocene to prepare ferrocenium tetrafluoroborate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nitrosonium tetrafluoroborate is a nitrosating agent and oxidizer used in organic synthesis. It is also used to convert arylamines directly to diazonium tetrafluoroborates, which undergo the Balz-Schiemann reaction in situ to give high yields of aryl fluorides. Further, it is used as a catalyst for iodination of aromatics by iodide ion with air as oxidant, conversion of secondary amines to nitrosamines and in deamination of primary amides. It is involved in the conversion of arylhydrazines to azides. In addition to this, it reacts with ferrocene to prepare ferrocenium tetrafluoroborate.

Solubility
Soluble in acetonitrile.

Notes
Hygroscopic. Air and moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids and amines.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14635-75-7
Molecular Formula BF4HNO2
Molecular Weight (g/mol) 133.82
MDL Number MFCD00011433
InChI Key XYRQURYIJKJVSF-UHFFFAOYSA-N
Synonym nitrosonium tetrafluoroborate, nitrosyl tetrafluoroborate, nitrosyltetrafluoroborate, nobf4, nitrilooxonium tetrafluoroborate, azanylidyneoxidanium tetrafluoroborate
PubChem CID 11137142
IUPAC Name azanylidyneoxidanium;tetrafluoroborate
SMILES [O-][NH+]=O.F[B-](F)(F)F

Specifications

Boiling Point 250°C (0.01 mmHg)(Sublimation)
Melting Point 250°C (Sublimation)
Quantity 2 g
UN Number UN3260
Merck Index 14,6649
Solubility Information Soluble in acetonitrile.
Formula Weight 116.82
Percent Purity 98%
Odor Irritating
Sensitivity Moisture sensitive
Density 2.185
Chemical Name or Material Nitrosonium tetrafluoroborate
Show More Show Less
compliance-icons
Product Identifier
  • Nitrosonium tetrafluoroborate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Nitrosonium tetrafluoroborate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.