missing translation for 'onlineSavingsMsg'
Learn More

Nitrotetrazolium Blue Chloride (Technical Grade), TRC

Product Code. 30469561
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30469561 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30469561 Supplier Toronto Research Chemicals Supplier No. TRCN572335250MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nitrotetrazolium Blue Chloride
CAS 298-83-9
Molecular Formula C40H30N10O6 • 2(Cl)
Quantity 250 mg
Synonym 3,3'-(3,3'-dimethoxy-4,4'-biphenylylene)bis[2-(p-nitrophenyl)-5-phenyl-2H-chloride] 2,2'-Bis(p-nitrophenyl)-5,5'-diphenyl-3,3'-(3,3'-dimethoxy-4,4'-biphenylylene)ditetrazolium chloride 2,2'-Bis(p-nitrophenyl)-5,5'-diphenyl-3,3'-(3,3'-dimethoxy-4,4'-diphenylene)ditetrazolium chloride 2,2'-Di-p-nitrophenyl-5,5'-diphenyl-3,3'-bis(3,3'-dimethoxy-4,4'-biphenylene)ditetrazolium chloride 2,2'-Dinitrophenyl-5,5'-diphenyl-3,3'-dimethoxy-4,4'-diphenylene)ditetrazolium chloride 3,3'-(3,3'-Dimethoxy-4,4'-biphenylylene)bis[2-(p-nitrophenyl)-5-phenyl-2H-tetrazolium chloride] 3,3'-(3,3'-Dimethoxy-4,4'-diphenylene)bis[2-(p-nitrophenyl)-5-phenyltetrazolium chloride] NBT NBT (dye) NSC 27622 NTB Nitro BT Nitro Blue Tetrazolium Nitro Blue Tetrazolium chloride Nitro TB Nitro Tetrazolium BT Nitrotetrazolium Chloride Blue Nitrotetrazolium blue Tetrazolium Nitro BT Tetrazolium nitro blue p-NBT p-Nitro blue tetrazolium p-Nitro blue tetrazolium chloride p-Nitrotetrazolium blue
InChI Formula InChI=1S/C40H30N10O6.2ClH/c1-55-37-25-29(13-23-35(37)47-43-39(27-9-5-3-6-10-27)41-45(47)31-15-19-33(20-16-31)49(51)52)30-14-24-36(38(26-30)56-2)48-44-40(28-11-7-4-8-12-28)42-46(48)32-17-21-34(22-18-32)50(53)54;;/h3-26H,1-2H3;2*1H/q+2;;/p-2
SMILES COC1=C(C=CC(=C1)C2=CC(=C(C=C2)[N+]3=NC(=NN3C4=CC=C(C=C4)[N+](=O)[O-])C5=CC=CC=C5)OC)[N+]6=NC(=NN6C7=CC=C(C=C7)[N+](=O)[O-])C8=CC=CC=C8.[Cl-].[Cl-]
IUPAC Name 2-[2-methoxy-4-[3-methoxy-4-[3-(4-nitrophenyl)-5-phenyltetrazol-2-ium-2-yl]phenyl]phenyl]-3-(4-nitrophenyl)-5-phenyltetrazol-2-ium;dichloride
Molecular Weight (g/mol) 746.7323545
Formula Weight 816.1727
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.