Learn More
Tocris Bioscience™ NK 252
GREENER_CHOICE
Nrf2 activator
Brand:  Tocris Bioscience™ 5293/10
Encompass
Product Code. 15706676
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
NK 252 is a Nrf2 activator; interacts with the Nrf2-binding site of Keap1. Downregulates the expression of fibrogenic genes and increases antioxidant effects in NASH rat models of liver disease.
Specifications
Specifications
NK 252
N-[5-(2-Furanyl)-1,3,4-oxadiazol-2-yl]-N'-(2-pyridinylmethyl)urea
Soluble to 100mM in 1eq. HCl and to 100mM in DMSO
Bioactive Small Molecule
1414963-82-8
10 mg
Powder
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Research Use Only. Not For Human or Veterinary Use