missing translation for 'onlineSavingsMsg'
Learn More

NM107, MedChemExpress

Product Code. 16493988
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16493988 10 mg 10mg
16444996 100 mg 100mg
16440347 50 mg 50mg
16416707 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16493988 Supplier Medchem Express Supplier No. HY10468
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress NM107 (2'-C-Methylcytidine) is an nucleoside inhibitor of the hepatitis C virus (HCV) NS5B polymerase, the EC50 of NM107 in the wild-type replicon cells is 1.85 μM.

NM107 (2'-C-Methylcytidine) is an nucleoside inhibitor of the hepatitis C virus (HCV) NS5B polymerase, the EC50 of NM107 in the wild-type replicon cells is 1.85 μM.

Specifications

Chemical Name or Material NM107
CAS 20724-73-6
Color White
Purity Grade Notes Research
Molecular Formula C10H15N3O5
Quantity 10 mg
Synonym 2'-C-Methylcytidine NM-107
Solubility Information DMSO : 100 mg/mL (388.74 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (194.37 mM)
SMILES NC(C=CN1[C@@H]2O[C@H](CO)[C@@H](O)[C@@]2(C)O)=NC1=O
Molecular Weight (g/mol) 257.24
Formula Weight 257.24
Percent Purity 98.9%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.