missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NMDA

Product Code. 30001055
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001055 500 mg 500mg
18085632 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 30001055 Supplier Tocris Bioscience™ Supplier No. 0114/500
  • ONLINE EXCLUSIVE PRICE
    547.00 €
    516.60 € / 500mg
    Save 30.40 €
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price

Selective NMDA agonist

Prototypic NMDA receptor agonist.

Caged version also available (Cat. No 2224).

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6384-92-5
Molecular Formula C5H9NO4

Specifications

Quantity 500 mg
Formula Weight 147.13
Purity >99%
Molecular Weight (g/mol) 147.13
Chemical Name or Material NMDA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.