missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Nocodazole

Product Code. 30001212
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001212 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001212 Supplier R&D Systems™ Supplier No. 1228/10
  • ONLINE EXCLUSIVE PRICE
    190.30 €
    170.10 € / 10mg
    Save 20.20 €
    10% Off
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Microtubule inhibitor

Microtubule inhibitor; inhibits mitosis. Also inhibits autophagosome-lysosome fusion. Enhances homology-directed repair (HDR) efficiency 9 to 31% (depending on cell cycle phase) and increases Cas9-mediated gene editing frequencies.
TRUSTED_SUSTAINABILITY

Specifications

CAS 31430-18-9
Quantity 10 mg
Product Line Tocris
Molecular Formula C14H11N3O3S
Formula Weight 301.32
Purity >95%
Molecular Weight (g/mol) 301.32
Chemical Name or Material Nocodazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.