missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Nocodazole

Product Code. 17140322
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17140322 50 mg 50mg
17130322 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17140322 Supplier STEMCELL Technologies Supplier No. 74074
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Microtubule polymerization inhibitor; Inhibits beta-tubulin

  • Nocodazole is an anti-mitotic agent that reversibly interferes with the polymerization of microtubules (De Brabander et al.) Nocodazole binds to beta-tubulin and disrupts microtubule assembly/disassembly dynamics, impairing formation of the metaphase spindles during the cell division cycle
  • This prevents mitosis by inducing a G2/M-phase arrest and induces apoptosis in tumor cells (Jordan et al.)
  • Synchronizes human pluripotent stem cells (hPSCs) in the G2/M cell cycle phase, thereby increasing the efficiency of homology-directed repair (HDR) in CRISPR-Cas9 genome editing (Lin et al.; Yang et al.)
  • Induces apoptosis in leukemic cells without harming T cells and/or mesenchymal stromal cells that were recovered from the same patients (Frezzato et al.)

Specifications

CAS 31430-18-9
Synonym NSC 238159; Oncodazole; R 17934
Quantity 50 mg
Molecular Formula C14H11N3O3S
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.