missing translation for 'onlineSavingsMsg'
Learn More

Nocodazole, 98%

Product Code. 10383912 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
Packaging:
Glass bottle
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10383912 50 mg Glass bottle 50mg
10762633 10 mg Glass bottle 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10383912 Supplier Thermo Scientific Acros Supplier No. 358240500
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 31430-18-9 | C14H11N3O3S | 301.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31430-18-9
Molecular Formula C14H11N3O3S
Molecular Weight (g/mol) 301.32
MDL Number MFCD00005588
InChI Key KYRVNWMVYQXFEU-UHFFFAOYSA-N
Synonym nocodazole, oncodazole, nocodazol, nocodazolum, nocidazole, nocodazole usan:inn, nocodazol inn-spanish, nocodazolum inn-latin, unii-sh1wy3r615, methyl 5-2-thenoyl-2-benzimidazolecarbamate
PubChem CID 4122
ChEBI CHEBI:34892
IUPAC Name methyl N-[6-(thiophene-2-carbonyl)-1H-benzimidazol-2-yl]carbamate
SMILES COC(=O)NC1=NC2=C(N1)C=C(C=C2)C(=O)C3=CC=CS3

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 300.0°C to 305.0°C
Color Brown-Orange to White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 50 mg
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in dmso,dmf,ethanol,chloroform,acetone
Formula Weight 301.32
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Nocodazole, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Nocodazole
Signal Word
  • Danger
Hazard Category
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H341-Suspected of causing genetic defects.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H361d-Suspected of damaging the unborn child.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.