missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Superior durability with enhanced walls and crush-resistant 'turrets' at the corners
- Easy handling with tabs to prevent stacked reservoirs from sticking together
- Graduated 5, 10 mL
Specifications
Specifications
| Certifications/Compliance | Certified DNase, RNase and Pyrogen Free |
| Color | White |
| Capacity (Metric) | 25 mL |
| Material | Polystyrene |
| Sterility | Non-sterile |
| Type | Reagent Reservoir |
| For Use With (Application) | Pipetting |
| Packaging | Bulk Wrap |
| Autoclavable | Not Autoclavable |
| Channels | 1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?