missing translation for 'onlineSavingsMsg'
Learn More

Nonadecanoic acid, 98+%

Product Code. 10138222 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10138222 1 g Glass bottle 1g
10114840 5 g Glass bottle 5g
10391124 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10138222 Supplier Thermo Scientific Acros Supplier No. 207190010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 646-30-0 | C19H38O2 | 298.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 646-30-0
Molecular Formula C19H38O2
Molecular Weight (g/mol) 298.51
MDL Number MFCD00002754
InChI Key ISYWECDDZWTKFF-UHFFFAOYSA-N
Synonym n-nonadecanoic acid, nonadecylic acid, unii-h6m3vyc62p, h6m3vyc62p, fatty acids, c16-22, n-nonadecanoate, n-nonadecylate, nonadecyclic acid, c16-c22 alkylcarboxylic acid
PubChem CID 12591
ChEBI CHEBI:39246
IUPAC Name nonadecanoic acid
SMILES CCCCCCCCCCCCCCCCCCC(=O)O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 68°C to 70°C
Color White
Boiling Point 297°C (100.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)17CO2H
Quantity 1 g
Beilstein 02, II, 368
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hot alcohol and ether
Formula Weight 298.51
Percent Purity 98+%
Physical Form Fine Powder or Shiny Flakes
Chemical Name or Material Nonadecanoic acid
Show More Show Less
compliance-icons
Product Identifier
  • Nonadecanoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.