missing translation for 'onlineSavingsMsg'
Learn More

Nonadecanoic acid, 98+%

Product Code. p-3779342 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 10138222

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 646-30-0 | C19H38O2 | 298.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 646-30-0
Molecular Formula C19H38O2
Molecular Weight (g/mol) 298.51
MDL Number MFCD00002754
InChI Key ISYWECDDZWTKFF-UHFFFAOYSA-N
Synonym n-nonadecanoic acid, nonadecylic acid, unii-h6m3vyc62p, h6m3vyc62p, fatty acids, c16-22, n-nonadecanoate, n-nonadecylate, nonadecyclic acid, c16-c22 alkylcarboxylic acid
PubChem CID 12591
ChEBI CHEBI:39246
IUPAC Name nonadecanoic acid
SMILES CCCCCCCCCCCCCCCCCCC(=O)O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 68°C to 70°C
Color White
Boiling Point 297°C (100.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)17CO2H
Quantity 1 g
Beilstein 02, II, 368
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hot alcohol and ether
Formula Weight 298.51
Percent Purity 98+%
Physical Form Fine Powder or Shiny Flakes
Chemical Name or Material Nonadecanoic acid
Show More Show Less
compliance-icons
Product Identifier
  • Nonadecanoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.