missing translation for 'onlineSavingsMsg'
Learn More

Nonafluoro-1-butanesulfonyl Fluoride, TRC

Product Code. 30473499
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30473499 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30473499 Supplier Toronto Research Chemicals Supplier No. TRCN6493201G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nonafluoro-1-butanesulfonyl Fluoride
CAS 375-72-4
Molecular Formula C4 F10 O2 S
Quantity 1 g
Synonym Perfluorobutanesulfonyl Fluoride, 1,1,2,2,3,3,4,4,4-Nonafluoro-1-butanesulfonyl fluoride (ACI), 1-Butanesulfonyl fluoride','nonafluoro- (6CI), 1,1,2,2,3,3,4,4,4-Nonafluorobutan-1-sulfonyl fluoride, 1,1,2,2,3,3,4,4,4-Nonafluorobutanesulfonyl fluoride, n-Nonafluorobutanesulfonyl fluoride, Nonafluoro-1-butanesulfonyl fluoride, Nonafluorobutanesulfonic acid fluoride, Nonafluorobutanesulfonyl fluoride, Nonaflyl fluoride, Perfluoro-1-butanesulfonyl fluoride, Perfluorobutan-1-sulfonyl fluoride, Perfluorobutanesulfonyl fluoride, Perfluorobutylsulfonyl fluoride, 1-Butanesulfonyl fluoride','1,1,2,2,3,3,4,4,4-nonafluoro- (8CI','9CI','ACI), 1,1,2,2,3,3,4,4,4-Nonafluoro-1-butanesulfonyl fluoride (ACI)
InChI Formula InChI=1S/C4F10O2S/c5-1(6,3(9,10)11)2(7,8)4(12,13)17(14,15)16
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)S(=O)(=O)F
IUPAC Name 1,1,2,2,3,3,4,4,4-nonafluorobutane-1-sulfonyl fluoride
Molecular Weight (g/mol) 302.09
Formula Weight 301.9459
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.