missing translation for 'onlineSavingsMsg'
Learn More

Nonafluoro-1-iodobutane, 97%

Product Code. 11452658
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11452658 100 g 100g
11432658 5 g 5g
11442658 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11452658 Supplier Thermo Scientific Alfa Aesar Supplier No. A16671.22
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 423-39-2 | C4F9I | 345.934 g/mol

Nonafluoro-1-iodobutane is used as iodine atom donor in the synthesis of iodocyclohexane. It is used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nonafluoro-1-iodobutane is used as iodine atom donor in the synthesis of iodocyclohexane. It is used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Not miscible in water.

Notes
Light Sensitive. Store in the dark away from oxidizing agents. Incompatible with alkali metals and light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 423-39-2
Molecular Formula C4F9I
Molecular Weight (g/mol) 345.934
MDL Number MFCD00001062
InChI Key PGRFXXCKHGIFSV-UHFFFAOYSA-N
Synonym perfluorobutyl iodide, nonafluoro-1-iodobutane, nonafluorobutyl iodide, perfluorobutyliodide, n-nonafluorobutyl iodide, perfluoro-n-butyl iodide, 1-iodoperfluorobutane, ccris 9008, 1-iodononafluorobutane, butane, 1,1,1,2,2,3,3,4,4-nonafluoro-4-iodo
PubChem CID 67917
IUPAC Name 1,1,1,2,2,3,3,4,4-nonafluoro-4-iodobutane
SMILES C(C(C(F)(F)I)(F)F)(C(F)(F)F)(F)F

Specifications

Melting Point -89°C to -87°C
Density 2.01
Boiling Point 67°C
Odor Pungent
Refractive Index 1.329
Quantity 100 g
UN Number UN3082
Beilstein 1777546
Sensitivity Light sensitive
Solubility Information Not miscible in water.
Formula Weight 345.93
Percent Purity 97%
Chemical Name or Material Nonafluoro-1-iodobutane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.