missing translation for 'onlineSavingsMsg'
Learn More

Nonafluorobutanesulfonyl chloride, 98%, Thermo Scientific Chemicals

Product Code. 11357279
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11357279 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11357279 Supplier Thermo Scientific Alfa Aesar Supplier No. H27437.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2991-84-6 | Molecular Formula: C4ClF9O2S | Molecular Weight: 318.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2991-84-6
Molecular Formula C4ClF9O2S
Molecular Weight (g/mol) 318.54
MDL Number MFCD03412262
InChI Key IRFCLLARAUQTNK-UHFFFAOYSA-N
Synonym nonafluoro-1-butanesulfonyl chloride, nonafluorobutane-1-sulfonyl chloride, perfluorobutanesulfonyl chloride, perfluorobutanesulphonyl chloride, nonafluorobutanesulfonyl chloride, 1-butanesulfonylchloride, 1,1,2,2,3,3,4,4,4-nonafluoro, acmc-20ak1o, perfluorobutylsulfonyl chloride, nonafluoro-n-butanesulfonyl chloride
PubChem CID 324955
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)S(Cl)(=O)=O

Specifications

Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
IUPAC Name 1,1,2,2,3,3,4,4,4-nonafluorobutane-1-sulfonyl chloride
Formula Weight 318.55
Percent Purity 98%
Chemical Name or Material Nonafluorobutanesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.