missing translation for 'onlineSavingsMsg'
Learn More

Nonanal, 97%

Product Code. 11428436
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11428436 100 g 100g
11438436 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11428436 Supplier Thermo Scientific Alfa Aesar Supplier No. A15908.22
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 124-19-6 | C9H18O | 142.242 g/mol

Nonanal is used for fragranced end-products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nonanal is used for fragranced end-products.

Solubility
Insoluble in water.

Notes
Incompatible with reducing agents, strong basesand strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-19-6
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.242
MDL Number MFCD00007030
InChI Key GYHFUZHODSMOHU-UHFFFAOYSA-N
Synonym pelargonaldehyde, 1-nonanal, nonanaldehyde, nonyl aldehyde, n-nonaldehyde, nonylic aldehyde, nonylaldehyde, nonaldehyde, n-nonanal, 1-nonaldehyde
PubChem CID 31289
ChEBI CHEBI:84268
IUPAC Name nonanal
SMILES CCCCCCCCC=O

Specifications

Density 0.825
Boiling Point 79°C to 81°C (12 mmHg)
Flash Point 63°C (145°F)
Refractive Index 1.424
Quantity 100 g
Beilstein 1236701
Solubility Information Insoluble in water.
Formula Weight 142.24
Percent Purity 97%
Chemical Name or Material Nonanal

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.