missing translation for 'onlineSavingsMsg'
Learn More

Nonanoic acid, 97%

Product Code. 11487164
Change view
Click to view available options
Quantity:
250 mL
1000 mL
Unit Size:
1000mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11487164 250 mL 250mL
11477164 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11487164 Supplier Thermo Scientific Alfa Aesar Supplier No. B21568.AK
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 112-05-0 | C9H18O2 | 158.241 g/mol

Nonanoic acid is used in the preparation of plasticizers and lacquers. It is commonly used in conjunction with glyphosate, for a quick burn-down effect in the control of weeds in turfgrass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nonanoic acid is used in the preparation of plasticizers and lacquers. It is commonly used in conjunction with glyphosate, for a quick burn-down effect in the control of weeds in turfgrass.

Solubility
Soluble in water at 20°C,0.26g/L. Soluble in alcohol and dipropylene glycol.

Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-05-0
Molecular Formula C9H18O2
Molecular Weight (g/mol) 158.241
MDL Number MFCD00004433
InChI Key FBUKVWPVBMHYJY-UHFFFAOYSA-N
Synonym pelargonic acid, n-nonanoic acid, nonylic acid, pelargic acid, nonoic acid, n-nonylic acid, n-nonoic acid, 1-octanecarboxylic acid, pelargon, cirrasol 185a
PubChem CID 8158
ChEBI CHEBI:29019
IUPAC Name nonanoic acid
SMILES CCCCCCCCC(=O)O

Specifications

Melting Point 10°C to 12°C
Density 0.905
pH 3.5
Boiling Point 253°C to 255°C
Flash Point 140°C (284°F)
Odor Unpleasant
Refractive Index 1.4315
Quantity 250 mL
UN Number UN3265
Beilstein 1752351
Merck Index 14,7070
Solubility Information Soluble in water at 20°C,0.26g/L. Soluble in alcohol and dipropylene glycol.
Formula Weight 158.24
Percent Purity 97%
Chemical Name or Material Nonanoic acid
Show More Show Less
compliance-icons
Product Identifier
  • Nonanoic acid
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Nonanoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.