missing translation for 'onlineSavingsMsg'
Learn More

Nonidet™ P40 Substitute, Ultrapure, Thermo Scientific Chemicals

Product Code. 15875388 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Plastic bottle
Unit Size:
1 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15875388 500 mL Plastic bottle 500mL
15885388 1 L Plastic bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15875388 Supplier Thermo Scientific Supplier No. J19628.AP
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Chemical Identifiers

CAS 9002-93-1
Molecular Formula C16H26O2
Molecular Weight (g/mol) 250.38
MDL Number MFCD00132505
InChI Key JYCQQPHGFMYQCF-UHFFFAOYSA-N
Synonym Igepal CA to 630,Polyethylene glycol tert to octylphenyl ether
PubChem CID 5590
IUPAC Name 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethanol
SMILES CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1

Specifications

Physical Form Viscous Liquid
Boiling Point >250°C
Melting Point ∼5°C
Density 1.061
Quantity 500 mL
Flash Point >110°C
Packaging Plastic bottle
Specific Gravity 1.061
Formula Weight ca 625
Chemical Name or Material Nonidet™ P40 Substitute

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.