missing translation for 'onlineSavingsMsg'
Learn More

Nonylmagnesium bromide, 1M solution in diethyl ether, AcroSeal™

Product Code. 11926021 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11926021 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11926021 Supplier Thermo Scientific Acros Supplier No. 440781000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 39691-62-8 | C9H19BrMg | 231.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 39691-62-8, 60-29-7
Color Colorless to Brown
Density 0.8490g/mL
Boiling Point 35.0°C
Flash Point −30°C
Packaging AcroSeal™ Glass Bottle
Molecular Formula C9H19BrMg
Linear Formula CH3(CH2)8MgBr
MDL Number MFCD03094048
Quantity 100 mL
Specific Gravity 0.849
Synonym nonylmagnesium bromide, nonylmagnesium bromide solution, ifqpktpqjlazhq-uhfffaoysa-m, nonylmagnesium bromide 1.0m solution i, nonylmagnesium bromide, 0.5m in tetrahydrofuran, nonylmagnesium bromide 1.0m solution i, nonylmagnesium bromide solution, 1.0 m in diethyl ether, grignard reagent
Solubility Information Solubility in water: reacts
InChI Key IFQPKTPQJLAZHQ-UHFFFAOYSA-M
SMILES CCCCCCCCC[Mg]Br
IUPAC Name magnesium;nonane;bromide
Molecular Weight (g/mol) 231.46
PubChem CID 11042600
Formula Weight 231.46
Physical Form Liquid
Chemical Name or Material Nonylmagnesium bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.