missing translation for 'onlineSavingsMsg'
Learn More

Norbixin, TRC

Product Code. 17968664
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17968664 50 mg 50mg
17947154 10 mg 10mg
17928774 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17968664 Supplier Toronto Research Chemicals Supplier No. TRCN66112050MG
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 542-40-5
Molecular Formula C24H28O4
Molecular Weight (g/mol) 380.48
Synonym 2,4,6,8,10,12,14,16,18-Eicosanonaenedioic acid, 4,8,13,17-tetramethyl-, (2E,4E,6E,8E,10E,12E,14E,16E,18E)-, 2,4,6,8,10,12,14,16,18-Eicosanonaenedioic acid, 4,8,13,17-tetramethyl- (8CI), 6,6'-Diapo-ψ,ψ-carotenedioic acid, Norbixin (6CI,7CI), (2E,4E,6E,8E,10E,12E,14E,16E,18E)-4,8,13,17-Tetramethyl-2,4,6,8,10,12,14,16,18-eicosanonaenedioic acid, AFC-WS 4600P, all-trans-Norbixin, β-Norbixin
IUPAC Name (2E,4E,6E,8E,10E,12E,14E,16E,18E)-4,8,13,17-tetramethylicosa-2,4,6,8,10,12,14,16,18-nonaenedioic acid
SMILES C\C(=C/C=C/C=C(\C)/C=C/C=C(\C)/C=C/C(=O)O)\C=C\C=C(/C)\C=C\C(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C24H28O4/c1-19(11-7-13-21(3)15-17-23(25)26)9-5-6-10-20(2)12-8-14-22(4)16-18-24(27)28/h5-18H,1-4H3,(H,25,26)(H,27,28)/b6-5+,11-7+,12-8+,17-15+,18-16+,19-9+,20-10+,21-13+,22-14+
Formula Weight 380.1988
Chemical Name or Material Norbixin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.