missing translation for 'onlineSavingsMsg'
Learn More

Norbornane-2-carboxylic acid, 98%, predominantly endo isomer

Product Code. 11308175
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11308175

Brand: Thermo Scientific Alfa Aesar L08439.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 824-62-4 | C8H12O2 | 140.18 g/mol

Norbornane-2-carboxylic acid is used in the preparation of 2-Bromonorbornane-1-carboxylic acid. It is also used as Pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Norbornane-2-carboxylic acid is used in the preparation of 2-Bromonorbornane-1-carboxylic acid. It is also used as Pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents and strong bases. Incompatible with oxidizing agents, Strong acids, Strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 824-62-4
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.18
MDL Number MFCD00167749
InChI Key JESWDXIHOJGWBP-UHFFFAOYNA-N
Synonym bicyclo 2.2.1 heptane-2-carboxylic acid, 2-norbornanecarboxylic acid, norbornane-2-carboxylic acid, 2-norbornanecarboxylic acid, endo, bicyclo 2.2.1 heptane-2-carboxylic acid, endo, exo-norbornane-2-carboxylic acid, bicyclo 2.2.1 heptane-3-carboxylic acid, 2-norbornanecarboxlic acid
PubChem CID 79113
SMILES OC(=O)C1CC2CCC1C2

Specifications

Melting Point ∼40°C
Boiling Point 134°C to 136°C (16 mmHg)
Flash Point >100°C (212°F)
Refractive Index 1.4855
Quantity 5 g
UN Number UN3261
Beilstein 2042631
Solubility Information Insoluble in water.
IUPAC Name bicyclo[2.2.1]heptane-2-carboxylic acid
Formula Weight 140.18
Percent Purity 98%
Chemical Name or Material Norbornane-2-carboxylic acid, predominantly endo isomer
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.