Learn More
Norbornene, 99%
Description
Norbornene is used as a monomer and as an intermediate in organic synthesis. It is also used to prepare pharmaceutical intermediates and specialty fragrances. It reacts with ethylene to get cyclic olefin copolymer. Further, it is used in transition metal catalysis to affect migration of electrophilic transition metals. It acts as a monomer in ring-opening metathesis polymerizations with the Grubbs' catalyst. Its polymer, polynorbornene is used in the rubber industry for anti-vibration, anti-impact in personal protective equipment and grip improvement in toy tires, racing tires and transmission system.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Norbornene is used as a monomer and as an intermediate in organic synthesis. It is also used to prepare pharmaceutical intermediates and specialty fragrances. It reacts with ethylene to get cyclic olefin copolymer. Further, it is used in transition metal catalysis to affect migration of electrophilic transition metals. It acts as a monomer in ring-opening metathesis polymerizations with the Grubbs′ catalyst. Its polymer, polynorbornene is used in the rubber industry for anti-vibration, anti-impact in personal protective equipment and grip improvement in toy tires, racing tires and transmission system.
Solubility
Soluble in ethanol, ether and most organic solvents. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 44°C to 47°C |
| Density | 0.955 |
| Boiling Point | 95°C to 96°C |
| Flash Point | −15°C (5°F) |
| Odor | Pungent |
| Quantity | 100 g |
| UN Number | UN1325 |
| Beilstein | 1098544 |
| Solubility Information | Soluble in ethanol,ether and most organic solvents. Insoluble in water. |
| Formula Weight | 94.16 |
| Show More |
Seguridad y manipulación
- Norbornene
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 2
- Flammable solid Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 3
- H228-Flammable solid.
- H304-May be fatal if swallowed and enters airways.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H361-Suspected of damaging fertility or the unborn child.
- H411-Toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P331-Do NOT induce vomiting.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contain: Bicyclo[2.2.1]hept-2-ene
RUO – Research Use Only
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.