missing translation for 'onlineSavingsMsg'
Learn More

Norbornylene, 99%, stabilized

Product Code. 10694982 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
Packaging:
Glass bottle
Unit Size:
25g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10694982 500 g Glass bottle 500g
10396670 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10694982 Supplier Thermo Scientific Acros Supplier No. 129245000
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 498-66-8 | C7H10 | 94.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 498-66-8
Molecular Formula C7H10
Molecular Weight (g/mol) 94.15
MDL Number MFCD00082304
InChI Key JFNLZVQOOSMTJK-UHFFFAOYSA-N
Synonym bicyclo 2.2.1 hept-2-ene, norbornylene, 2-norbornene, norbornene, norcamphene, norfenchene, 2-norbornylene, 3,6-endomethylenecyclohexene, bicyclo 2.2.1-2-heptene, bicyclo 2.2.1.-2-heptene
PubChem CID 10352
IUPAC Name bicyclo[2.2.1]hept-2-ene
SMILES C1CC2CC1C=C2

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 42.0°C to 46.0°C
Color White
Boiling Point 96.0°C
Flash Point −15°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 500 g
Fieser 01,757; 02,300
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in toluene and benzene
Stabilizer 0.1% phenolic antioxidant
Formula Weight 94.15
Percent Purity 99%
Physical Form Crystalline Mass
Chemical Name or Material Norbornylene, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.