missing translation for 'onlineSavingsMsg'
Learn More

Norbornylene, 99%, stabilized

Product Code. 15364968 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
2.5 kg
Unit Size:
2.5kg
This item is not returnable. View return policy

Product Code. 15364968

Brand: Thermo Scientific Acros 129240025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 498-66-8 | C7H10 | 94.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 498-66-8
Melting Point 42.0°C to 46.0°C
Boiling Point 96.0°C
Flash Point −15°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Molecular Formula C7H10
Quantity 2.5 kg
Fieser 01,757; 02,3
Synonym bicyclo 2.2.1 hept-2-ene, norbornylene, 2-norbornene, norbornene, norcamphene, norfenchene, 2-norbornylene, 3,6-endomethylenecyclohexene, bicyclo 2.2.1-2-heptene, bicyclo 2.2.1.-2-heptene
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in toluene and benzene
InChI Key JFNLZVQOOSMTJK-UHFFFAOYSA-N
SMILES C1CC2CC1C=C2
IUPAC Name bicyclo[2.2.1]hept-2-ene
Molecular Weight (g/mol) 94.15
PubChem CID 10352
Formula Weight 94.15
Percent Purity 99%
Chemical Name or Material Norbornylene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.