missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Norethindrone acetate, MedChemExpress
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Description
Norethindrone acetate is a female hormone used for the research of endometriosis.
Specifications
Specifications
| Chemical Name or Material | Norethindrone acetate |
| CAS | 51-98-9 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C22H28O3 |
| Quantity | 100 mg |
| Synonym | 19-Norethindrone acetate |
| Solubility Information | DMSO : ≥ 100 mg/mL (293.72 mM) ∣H2O : 0.67 mg/mL (1.97 mM; Need ultrasonic) |
| SMILES | C#C[C@]1(OC(C)=O)CC[C@@]2([H])[C@]3([H])CCC4=CC(CC[C@]4([H])[C@@]3([H])CC[C@]12C)=O |
| Molecular Weight (g/mol) | 340.46 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction