missing translation for 'onlineSavingsMsg'
Learn More

Norfloxacin, TRC

Product Code. 17982355
Change view
Click to view available options
Quantity:
50 mg
1 g
5 g
Unit Size:
1g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17982355 1 g 1g
17972695 5 g 5g
17912035 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17982355 Supplier Toronto Research Chemicals Supplier No. TRCN6810001G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70458-96-7
Molecular Formula C16 H18 F N3 O3
Molecular Weight (g/mol) 319.33
Synonym 3-Quinolinecarboxylic acid, 1-ethyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-, 1-Ethyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic acid, 1-Ethyl-6-fluoro-1,4-dihydro-7-(1-piperazinyl)-4-oxo-3-quinolinecarboxylic acid, 4803P, AM 715, Amicrobin, Baccidal, Barazan, Barotham, Chibroxin, Chibroxine, Chibroxol, Esclebin, Floxacin, Floxacin 400, Fulgram, Gonorcin, Lexinor, Mk 0366, N-Demethylpefloxacin, N-Desmethylpefloxacin, Neofloxacin, Neofloxin, Noflo, Nofocin, Nolicin, Noracin, Noraxin, Norbid, Norflox, Norflox 400, Norfloxacin, Norfloxacin Bayvit, Norfloxacine, Norfloxacine EG, Norfloxacine Ratiopharm, Norfloxacine Teva, Norfloxin, Norilet, Normax, Norocin, Noroxacine, Noroxin, Noroxine, Norphloxacine, Norxacin, Renosept, Sebercim, Spectrama, Urekacin, Uriflox, Uroflox, Urotrobel, Uroxacin, Utin, Utinor, Zoroxin
IUPAC Name 1-ethyl-6-fluoro-4-oxo-7-piperazin-1-ylquinoline-3-carboxylic acid
SMILES CCN1C=C(C(=O)O)C(=O)c2cc(F)c(cc12)N3CCNCC3

Specifications

Quantity 1 g
InChI Formula InChI=1S/C16H18FN3O3/c1-2-19-9-11(16(22)23)15(21)10-7-12(17)14(8-13(10)19)20-5-3-18-4-6-20/h7-9,18H,2-6H2,1H3,(H,22,23)
Formula Weight 319.1332
Percent Purity >95%
Chemical Name or Material Norfloxacin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.