missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Norite from Stillwater Complex, MT, CRM, ARMI∣MBH
Description
Norite from Stillwater Complex, MT, CRM
Specifications
Specifications
| Chemical Name or Material | Phosphorus/Cerium/Neodymium/Lead/Niobium/Ytterbium/Hafnium/Praseodymium/Thorium/Holmium/Uranium/Lutetium/Terbium/Thulium/Titanium/Potassium/Manganese/Potassium oxide/Manganese(II) oxide/Titanium dioxide/Samarium/Europium/Erbium/Gadolinium/Dysprosium/Lanthanum/Loss on ignition/Rubidium/Sodium/Sodium oxide/Yttrium/Lithium/Iron/Zirconium/Total Iron expressed as Fe2O3/Magnesium/Magnesium oxide/Calcium/Scandium/Copper/Gallium/Aluminium/Calcium oxide/Barium/Zinc/Cobalt/Silicon/Aluminium oxide/Vanadium/Silicon dioxide/Strontium/Nickel/Chromium |
| CAS | 14808-60-7 |
| Quantity | 75 g |
| Form | Fine Powder |
| Concentration or Composition (by Analyte or Components) | Phosphorus (30) mg/kg/Cerium (1) mg/kg/Neodymium (0.5) mg/kg/Lead (0.3) mg/kg/Niobium (0.2) mg/kg/Ytterbium (0.2) mg/kg/Hafnium (0.1) mg/kg/Praseodymium (0.1) mg/kg/Thorium (0.09) mg/kg/Holmium (0.05) mg/kg/Uranium (0.04) mg/kg/Lutetium (0.03) mg/kg/Terbium (0.03) mg/kg/Thulium (0.03) mg/kg/Titanium 0.038 %/Potassium 0.041 %/Manganese 0.046 %/Potassium oxide 0.049 %/Manganese(II) oxide 0.059 %/Titanium dioxide 0.064 %/Samarium 0.13 mg/kg/Europium 0.134 mg/kg/Erbium 0.18 mg/kg/Gadolinium 0.19 mg/kg/Dysprosium 0.22 mg/kg/Lanthanum 0.52 mg/kg/Loss on ignition 0.78 %/Rubidium 0.86 mg/kg/Sodium 0.91 %/Sodium oxide 1.23 %/Yttrium 1.37 mg/kg/Lithium 1.4 mg/kg/Iron 2.34 %/Zirconium 2.8 mg/kg/Total Iron expressed as Fe2O3 3.34 %/Magnesium 4.08 %/Magnesium oxide 6.77 %/Calcium 10 %/Scandium 11 mg/kg/Copper 11.4 mg/kg/Gallium 12.1 mg/kg/Aluminium 13.4 %/Calcium oxide 14 %/Barium 16.3 mg/kg/Zinc 17.9 mg/kg/Cobalt 21.8 mg/kg/Silicon 22.5 %/Aluminium oxide 25.4 %/Vanadium 44 mg/kg/Silicon dioxide 48.1 %/Strontium 97 mg/kg/Nickel 158 mg/kg/Chromium 586 mg/kg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?