missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Normal Goat Serum (10%)

Product Code. 10777043
Change view
Click to view available options
Quantity:
15 mL
100 mL
Unit Size:
100mL
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10777043 15 mL Each
10120363 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10777043 Supplier Invitrogen™ Supplier No. 50197Z
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Normal Goat Serum (10%)

This 10% Normal Goat Serum blocking solution is ready-to-use, in phosphate buffered saline (pH 7.4) with 0.1% sodium azide. This normal goat serum is intended for use as a blocking solution when a second antibody of goat origin is applied in an immunohistochemical staining procedure.

To block non-specific binding from the goat second antibody, incubate the tissue section with 10% Normal Goat Serum for 10 minutes at room temperature prior to the primary antibody incubation step.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 2-8°C
Species Goat/Caprine
Quantity 15 mL
Shipping Condition Wet Ice

For Laboratory Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.