missing translation for 'onlineSavingsMsg'
Learn More

Normal Goat Serum (10%)

Product Code. p-7076119
Click to view available options
Quantity:
15 mL
100 mL
Unit Size:
100mL
Each
This item is not returnable. View return policy

Product Code. 10777043

Brand: Invitrogen 50197Z

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Normal Goat Serum (10%)

This 10% Normal Goat Serum blocking solution is ready-to-use, in phosphate buffered saline (pH 7.4) with 0.1% sodium azide. This normal goat serum is intended for use as a blocking solution when a second antibody of goat origin is applied in an immunohistochemical staining procedure.

To block non-specific binding from the goat second antibody, incubate the tissue section with 10% Normal Goat Serum for 10 minutes at room temperature prior to the primary antibody incubation step.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 2-8°C
Species Goat/Caprine
Quantity 15 mL
Shipping Condition Wet Ice

For Laboratory Use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.