missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Normal Human Serum

Product Code. 10406144
Change view
Click to view available options
Quantity:
120 mg
Unit Size:
2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10406144 120 mg 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10406144 Supplier Invitrogen™ Supplier No. 31876
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Human Control

For blocking purposes, use 5% (v/v) solution (1:20 dilution from rehydrated volume). Rehydration according to instruction yields 100% serum. Restoration and Storage: Store product at 4°C until opened. Restore with 2.0 mL distilled water. Centrifuge product if it is not completely clear after standing for 1-2 hours at room temperature. Product may be stored for up to several weeks at 4°C as an undiluted liquid. After dilution, do not use for more than one day. To extend the shelf-life of this product, freeze rehydrated undiluted product in small aliquots at -20°C. Country of Origin: USA.

Normal Sera are non-immune serum samples from goat, human, mouse, rabbit and other species that contain the usual complement of serum proteins, including the various classes of immunoglobulins that exist in healthy animals of the particular species. Normal serum provides a sufficient quantity of endogenous proteins to saturate and block nonspecific binding sites on fixed samples or assay substrates. When used as an antibody diluent, non-immune serum also supplies a native chemical environment for antibodies to function properly without adding foreign protein components. Normal serum is frequently used for blocking or saturating generalized binding interactions for immunodetection methods, especially those involving tissue samples such as immunohistochemistry (IHC). Normal sera are also useful as controls for testing general and specific antibody purification methods.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 4°C
Form Lyophilized
Product Type Normal Human Serum
Quantity 120 mg
Concentration 60 mg/mL
Formulation Whole serum, PBS with 0.05% sodium azide; pH 7.6
For Use With (Application) Blocking Assay, Control, Immunohistochemistry, Immunocytochemistry
Isotype Ig Mix
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.