Learn More
Invitrogen™ Normal Mouse Serum
Descripción
For blocking purposes, use 5% (v/v) solution (1:20 dilution from rehydrated volume). Rehydration according to instruction yields 100% serum. Restoration and Storage: Store product at 4°C until opened. Restore with 2.0 mL distilled water. Centrifuge product if it is not completely clear after standing for 1-2 hours at room temperature. Product may be stored for up to several weeks at 4°C as an undiluted liquid. After dilution, do not use for more than one day. To extend the shelf-life of this product, freeze rehydrated undiluted product in small aliquots at 20°C. Country of Origin: USA.
Especificaciones
Especificaciones
| Content And Storage | 4°C |
| Form | Lyophilized |
| Product Type | Normal Mouse Serum |
| Quantity | 2 mL |
| Concentration | 50.0 mg/mL |
| Formulation | Whole serum, PBS with 0.05% sodium azide; pH 7.6 |
| For Use With (Application) | Control, Immunocytochemistry, Immunohistochemistry, Blocking Assay |
| Isotype | Ig |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.