missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Normal Mouse Serum

Product Code. 10140073
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10140073 10 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10140073 Supplier Invitrogen™ Supplier No. 10410
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Mouse Control

If the reagent is being diluted, it is recommended that only the quantity to be used within one week be diluted.

Normal Sera are non-immune serum samples from goat, human, mouse, rabbit and other species that contain the usual complement of serum proteins, including the various classes of immunoglobulins that exist in healthy animals of the particular species. Normal serum provides a sufficient quantity of endogenous proteins to saturate and block nonspecific binding sites on fixed samples or assay substrates. When used as an antibody diluent, non-immune serum also supplies a native chemical environment for antibodies to function properly without adding foreign protein components. Normal serum is frequently used for blocking or saturating generalized binding interactions for immunodetection methods, especially those involving tissue samples such as immunohistochemistry (IHC). Normal sera are also useful as controls for testing general and specific antibody purification methods.
TRUSTED_SUSTAINABILITY

Spezifikation

Color Amber, Clear
Content And Storage -20°C
Description Normal Mouse Serum Control, 10mL, Amber/Clear, Liquid, Mouse Species Reactivity, Mouse Immunoglobulins Immunogen, Unconjugated, 0.2μm Filtration, Whole Serum Storage Buffer, Contains 0.1% Sodium Azide as Preservative, –20°C Storage Temperature
Form Liquid
Product Type Mouse Serum
Quantity 10 mL
Concentration Conc. Not Determined
Formulation Whole serum with 0.1% sodium azide
For Use With (Application) Control
Preservative 0.1% Sodium azide
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.