missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ North™ Finger Cots

Product Code. 11746574 Shop All Cole-Parmer Products
Change view
Click to view available options
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11746574 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11746574 Supplier Cole-Parmer™ Supplier No. 8160552

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Finger cots provide effective protection for delicate applications

  • Rolled edges ensure quick and easy donning

Specifications

Certifications/Compliance ASTM D3772
Color Clear
Material Latex
Size Medium
For Use With (Application) Use when Sensitivity and Dexterity are needed
Quantity 1440Pack
Length (Metric) 70 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.