missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ North2South™ Chemiluminescent Hybridization and Detection Kit

Product Code. 11869320 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
375 mL
Unit Size:
375mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11869320 375 mL 375mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11869320 Supplier Thermo Scientific™ Supplier No. 37555

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Acquire consistent, highly sensitive Southern blot results with a novel ECL HRP substrate and optimized hybridization and blocking buffers.

Thermo Scientific™ North2South Chemiluminescent Detection Kit combines a novel ECL substrate for horseradish peroxidase (HRP) and optimized hybridization and blocking buffers to ensure consistent, high-sensitivity Southern blot results.

The Thermo Scientific North2South Hybridization and Detection Kit combines a novel enhanced luminol substrate for horseradish peroxidase (HRP) with optimized pre-made hybridization and blocking buffers to ensure consistent results and high sensitivity. Any nucleic acid blot can be hybridized with the appropriate biotinylated (biotin-labeled) oligonucleotide probe. After a brief membrane blocking step, the biotin label is detected with streptavidin-HRP and a 5-minute incubation with the chemiluminescent substrate. Results are easily recorded by brief exposure to X-ray film (1-15 minutes) or CCD camera.

Highlights:

  • Sensitivity equal to or greater than 32P
  • Amenable to many formats
  • High level of light output and consistent low background allows for very short exposure times using cooled CCD camera systems or film
  • Ready-to-use buffers and short process time makes switching from radioactive detection to chemiluminescent detection an easy transition, even for the novice user
TRUSTED_SUSTAINABILITY

Specifications

Formulation Yields 60mM sodium citrate, 600mM sodium chloride, 0.2% SDS, pH 7.9 when diluted 1:1 in water
For Use With (Application) Non-radioactive Southern and Northern blotting
Product Type North2South Hybridization Stringency Wash Buffer (2X)
Quantity 375 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.