missing translation for 'onlineSavingsMsg'
Learn More

Nortriptyline hydrochloride, MedChemExpress

Product Code. 16454597
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16454597 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16454597 Supplier Medchem Express Supplier No. HYB1417
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Nortriptyline hydrochloride (Desmethylamitriptyline hydrochloride), the main active metabolite of Amitriptyline, is a tricyclic antidepressant used to relieve the symptoms of depression. Nortriptyline hydrochloride is a potent autophagy inhibitor.

Nortriptyline hydrochloride (Desmethylamitriptyline hydrochloride), the main active metabolite of Amitriptyline, is a tricyclic antidepressant used to relieve the symptoms of depression. Nortriptyline hydrochloride is a potent autophagy inhibitor.

Specifications

Chemical Name or Material Nortriptyline hydrochloride
CAS 894-71-3
Color White
Purity Grade Notes Research
Molecular Formula C19H22ClN
Quantity 50 mg
Synonym Desmethylamitriptyline hydrochlorideDesitriptilina hydrochloride
Solubility Information DMSO : 83.33 mg/mL (277.91 mM; Need ultrasonic) ∣H2O : 7.14 mg/mL (23.81 mM; Need ultrasonic)
SMILES CNCC/C=C1C2=CC=CC=C2CCC3=CC=CC=C\13.[H]Cl
Molecular Weight (g/mol) 299.84
Formula Weight 299.84
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.