Learn More
- Home
- Products
- Protein Analysis Reagents
- Crosslinking, Labeling and Protein Modification
- Protein Labeling Reagents
- Invitrogen™ NovaFluor™ Antibody Conjugation Kits
Invitrogen™ NovaFluor™ Antibody Conjugation Kits
Product Code. 31759085| Product Code. | Label or Dye | Product Type | Emission | Quantity | unitSize |
|---|---|---|---|---|---|
| 31759085 | NovaFluor™ Blue 745 | Conjugation Kit | 748 | 100 μg | 100µg |
| 17728303 | NovaFluor™ Blue 510 | Conjugation Kit | 510 nm | 100 μg | 100µg |
| 17768293 | NovaFluor™ Blue 530 | Conjugation Kit | 530 nm | 100 μg | 100µg |
| 17585325 | NovaFluor™ Blue 555 | Conjugation Kit | 555 nm | 100 μg | 100µg |
| 17738303 | NovaFluor™ Blue 585 | Conjugation Kit | 585 nm | 100 μg | 100µg |
| 17778293 | NovaFluor™ Blue 610/30S | Conjugation Kit | 610 nm | 100 μg | 100µg |
| 17748303 | NovaFluor™ Blue 610/70S | Conjugation Kit | 610 nm | 100 μg | 100µg |
| 17788293 | NovaFluor™ Blue 660/40S | Conjugation Kit | 660 nm | 100 μg | 100µg |
| 17826781 | NovaFluor™ Blue 690 | Conjugation Kit | 690 nm | 100 μg | 100µg |
| 17758303 | NovaFluor™ Blue 660/120S | Conjugation Kit | 660 nm | 100 μg | 100µg |
| 17708313 | NovaFluor™ Red 660 | Conjugation Kit | 660 nm | 100 μg | 100µg |
| 17718313 | NovaFluor™ Red 685 | Conjugation Kit | 685 nm | 100 μg | 100µg |
| 17728313 | NovaFluor™ Red 700 | Conjugation Kit | 700 nm | 100 μg | 100µg |
| 17738313 | NovaFluor™ Red 710 | Conjugation Kit | 710 nm | 100 μg | 100µg |
| 17846781 | NovaFluor™ Red 725 | Conjugation Kit | 725 nm | 100 μg | 100µg |
| 17856781 | NovaFluor™ Red 755 | Conjugation Kit | 755 nm | 100 μg | 100µg |
| 17798293 | NovaFluor™ Yellow 570 | Conjugation Kit | 570 nm | 100 μg | 100µg |
| 17768303 | NovaFluor™ Yellow 590 | Conjugation Kit | 590 nm | 100 μg | 100µg |
| 17708303 | NovaFluor™ Yellow 610 | Conjugation Kit | 610 nm | 100 μg | 100µg |
| 17718303 | NovaFluor™ Yellow 660 | Conjugation Kit | 660 nm | 100 μg | 100µg |
| 17778303 | NovaFluor™ Yellow 690 | Conjugation Kit | 690 nm | 100 μg | 100µg |
| 17788303 | NovaFluor™ Yellow 700 | Conjugation Kit | 700 nm | 100 μg | 100µg |
| 17798303 | NovaFluor™ Yellow 730 | Conjugation Kit | 730 nm | 100 μg | 100µg |
| 17836781 | NovaFluor™ Yellow 755 | Conjugation Kit | 755 nm | 100 μg | 100µg |
| 31759080 | NovaFluor™ Red 800 | Conjugation Kit | 799 | 100 μg | 100µg |
Description
NovaFluor Conjugation Kits provide a simple workflow to conjugate a blue, yellow-green or red excitable fluorophore to your antibody of choice in just 3 easy steps. The fluorescence-by-design Phiton platform enables the addition of parameters to your current panels in discovery research and resolves stained populations with near-zero cross excitation, with a lack of emission in the violet channels that plague other fluorophores.
NovaFluor Conjugation Kit provides features including:
- A clean and bright antibody label to increase the number of parameters on conventional and spectral flow cytometers
- Resolve populations with limited/near-zero cross excitation and the lowest spread performance of any blue, yellow-green or red excited fluorescent labels
- Simple 3-step protocol, with 60 sec. of hands-on time for conjugating 10–100 μg of antibody per label
- Up to 70% less excitation when compared to PE tandems, resulting in decreased spillover and higher resolution
3-step conjugation method
- Attach NovaFluor Linker to antibody of choice
- Remove excess NovaFluor Linker from DNA-conjugate antibody
- Anneal NovaFluor to oligo-conjugated antibody
Specifications
Specifications
| For Use With (Application) | Flow Cytometry |
| Quantity | 100 μg |
| Product Type | Conjugation Kit |
| Flow Cytometer Laser Lines | Blue |
| Label or Dye | NovaFluor™ Blue 745 |
| Label Type | Other Labels or Dyes |
| Color | Far-red |
| Product Line | NovaFluor |
| Emission | 748 |
| Excitation | 492 |
| Excitation/Emission | 492/748 |
| Show More Show Less |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.