Learn More
Merck Novagen™, Biotinylated Thrombin
Product Code. 17817547
Click to view available options
Quantity:
50 U
Unit Size:
Pack of 50
Product Code. 17817547
Brand: Merck 696723
Removes enzyme from cleavage reactions using immobilized streptavidin.
  • Covalently attached biotin
  • CAS: Ethylenediaminetetraacetic Acid: 60-00-4, Tris Base: 77-86-1
  • Packaging: Glass Bottle
TRUSTED_SUSTAINABILITY
Specifications
Quantity 50 U