missing translation for 'onlineSavingsMsg'
Learn More

Merck Novagen™ Pellet Paint™ NF Co-Precipitant

Product Code. 17937160
Click to view available options
No. of Reactions:
125
Unit Size:
Pack of 125
This item is not returnable. View return policy

Product Code. 17937160

Brand: Merck 707483

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Useful substitute for the original Pellet Paint Co-Precipitant in applications where fluorescent detection is used

  • Efficient and rapid precipitation of BigDye™ cycle sequencing products
  • Efficient removal of dye terminators
  • Direct visualization and tracking of precipitated material
  • No effect on sequencing reaction
  • Substitute for original Pellet Paint™ Co-Precipitant for fluorescent detection applications
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at −20°C
Label or Dye Non-fluorescent
No. of Reactions 125
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.