missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Novagen™ Pellet Paint™ NF Co-Precipitant
Click to view available options
No. of Reactions:
125
Unit Size:
Pack of 125
Description
- Efficient and rapid precipitation of BigDye™ cycle sequencing products
- Efficient removal of dye terminators
- Direct visualization and tracking of precipitated material
- No effect on sequencing reaction
- Substitute for original Pellet Paint™ Co-Precipitant for fluorescent detection applications
Specifications
Specifications
| Content And Storage | Store at −20°C |
| Label or Dye | Non-fluorescent |
| No. of Reactions | 125 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction