missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ Tris-Glycine Mini Protein Gels, 10%, 1.0 mm, 2D-well

Product Code. 12015715
Click to view available options
Quantity:
10 gels (1 box)
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 12015715

Brand: Invitrogen™ EC6076BOX

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Catalog numbers for gels refer to a single box. One box contains 10 gels.

Separation Range: 30 to 250 kd; Separation Type: Denaturing, Native; For Use With: Mini Gel Tank, XCell SureLock Mini

  • Do not contain SDS and can therefore be used to accurately separate both native and denatured protein
  • Provide reproducible separation of a wide range of proteins into well-resolved bands
  • Made with high-purity, strictly quality-controlled reagents: Tris base, HCl, acrylamide, bisacrylamide, TEMED, APS, and highly purified water

2D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage One box contains 10 gels. Store in refrigerator (2–8°C). Do not freeze. Shelf life is 8 weeks.
For Use With (Equipment) Mini Gel Tank, XCell SureLock Mini-Cell
Gel Percentage 10%
Gel Size Mini
Gel Thickness 1.0 mm
Gel Type Tris-Glycine
Product Type Homogeneous Page
Separation of Protein
Separation Range 30 to 250 kDa
Separation Type Denaturing, Native
Wells 1-well
Mode of Separation Molecular Weight
Product Line Novex
Quantity 10 gels (1 box)
Shipping Condition Wet Ice
System Type ZOOM™
Thickness 1.0 mm
Well Design 2D Well
Length (Metric) 8 cm
Length (Metric) Gel 8 cm
Width (Metric) 8 cm
Width (Metric) Gel 8 cm
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.