missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 10X Bolt™ Sample Reducing Agent

Product Code. p-4931808
Click to view available options
Quantity:
250 μL
10 mL
Unit Size:
10mL
250µL
This item is not returnable. View return policy

Product Code. 13296499

Brand: Invitrogen™ B0009

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

500mM dithiothreitol (DTT)

Optimized for use with Bolt™ Bis-Tris Plus gels, and available in a variety of formats.

  • Bolt™ Bis-Tris Plus gels are precast polyacrylamide gels designed for optimal separation of small- to medium-sized proteins under denaturing conditions
  • Similar to NuPAGE™ Bis-Tris gels, Bolt™ Bis-Tris Plus gels are designed to deliver consistent gel performance and provide a neutral pH environment that minimizes protein modifications
  • Bolt™ gels are ideal for western blot transfer and analysis, and any other techniques where protein integrity is crucial
  • Also use Bolt™ gels to obtain optimal results for day-to-day protein separation needs
  • Gels come in four gel types and multiple well formats
  • A Mini Gel Tank is required for use of all Bolt™ gels
  • Run times may vary depending upon the type of running buffer used
  • Store at 2°C to 8°C
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Sample Reducing Agent (10X) contains 500 mM dithiothreitol (DTT).

Store at 2°C to 8°C.
Concentration 10X, 500 mM
For Use With (Equipment) Bolt Mini Gel Tank
Product Type Sample Reducing Agent
Form Liquid
Formulation 500 mM dithiothreitol (DTT)
Gel Compatibility Bolt™ Bis-Tris Plus Gels
Product Line Bolt
Quantity 10 mL
Reagent Type Reducing Agent

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.